Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199732.2 | NP_001186661.1 | 2 - 3 | 224 | 70 | |
| GenBank mRNA | CX601235.1 | - | 2 - 3 | 247 | 70 | |
| CX601382.1 | - | 2 - 3 | 154 | 70 | ||
More Information
Set Membership: |
|

SELENOS
Genomic Map