Assay Details
| Gene Symbol: SELENOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199732.2 | NP_001186661.1 | 3 - 4 | 324 | 105 | |
| GenBank mRNA | CX601382.1 | - | 3 - 4 | 254 | 105 | |
| JL624734.1 | - | 1 - 2 | 97 | 105 | ||
More Information
Set Membership: |
|

SELENOS
Genomic Map