Assay Details
| Gene Symbol: PLA2G4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204234.1 | NP_001191163.1 | 3 - 4 | 354 | 69 | |
| XM_014733464.2 | XP_014588950.1 | 1367 | 69 | |||
More Information
Set Membership: |
|

PLA2G4B
Genomic Map