Assay Details
| Gene Symbol: NME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204742.1 | NP_001191671.1 | 2 - 3 | 191 | 65 | |
| XM_005597033.3 | XP_005597090.1 | 607 | 65 | |||
| XM_023651651.1 | XP_023507419.1 | 282 | 65 | |||
| GenBank mRNA | DN511274.1 | - | 2 - 3 | 220 | 65 | |
More Information
Set Membership: |
|

NME1
Genomic Map