Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204844.2 | NP_001191773.1 | 3 - 4 | 2239 | 86 | |
| XM_005599776.3 | XP_005599833.1 | 2335 | 86 | |||
More Information
Set Membership: |
|

STON1
Genomic Map