Assay Details
| Gene Symbol: LY49E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081829.2 | NP_001075298.1 | 2 - 3 | 223 | 112 | |
| XM_005610807.3 | XP_005610864.1 | 290 | 112 | |||
| GenBank mRNA | AB120389.1 | - | 2 - 3 | 190 | 112 | |
| AB120390.1 | - | 2 - 3 | 226 | 112 | ||
More Information
Set Membership: |
|

LY49E
Genomic Map