Assay Details
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081858.2 | NP_001075327.2 | 2 - 3 | 307 | 96 | |
| GenBank mRNA | AF372660.1 | - | 2 - 3 | 307 | 96 | |
| Gene Symbol: SCGB1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653827.1 | XP_023509595.1 | 96 | |||
More Information
Set Membership: |
|

SCGB1A1
Genomic Map