Assay Details
| Gene Symbol: APOBEC3Z2A-Z2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163681.1 | NP_001157153.1 | 4 - 5 | 543 | 126 | |
| GenBank mRNA | FJ174662.1 | - | 4 - 5 | 543 | 126 | |
| FJ527822.1 | - | 4 - 5 | 543 | 126 | ||
More Information
Set Membership: |
|

APOBEC3Z2A-Z2
Genomic Map