Assay Details
| Gene Symbol: PTGES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081935.1 | NP_001075404.1 | 3 | 1319 | 100 | |
| XM_023628603.1 | XP_023484371.1 | 1645 | 100 | |||
| GenBank mRNA | AY057096.1 | - | 3 | 1319 | 100 | |
More Information
Set Membership: |
|

PTGES
Genomic Map