Assay Details
| Gene Symbol: SPI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114533.1 | NP_001108005.1 | 4 | 1216 | 107 | |
| XM_023627719.1 | XP_023483487.1 | 1480 | 107 | |||
More Information
Set Membership: |
|

SPI2
Genomic Map