Assay Details
| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504287.5 | XP_001504337.1 | 493 | 84 | ||
| XM_005597648.3 | XP_005597705.1 | 452 | 84 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YWHAE| Gene Symbol: YWHAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504287.5 | XP_001504337.1 | 493 | 84 | ||
| XM_005597648.3 | XP_005597705.1 | 452 | 84 | |||
Set Membership: |
|