Assay Details
| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503661.6 | XP_001503711.1 | 702 | 98 | ||
| XM_023650706.1 | XP_023506474.1 | 692 | 98 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TENT5A| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503661.6 | XP_001503711.1 | 702 | 98 | ||
| XM_023650706.1 | XP_023506474.1 | 692 | 98 | |||
Set Membership: |
|