Assay Details
| Gene Symbol: EIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362563.4 | XP_003362611.1 | 350 | 97 | ||
| GenBank mRNA | JL622774.1 | - | 1 - 2 | 138 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362563.4 | XP_003362611.1 | 350 | 97 | ||
| GenBank mRNA | JL622774.1 | - | 1 - 2 | 138 | 97 | |
Set Membership: |
|