Assay Details
Gene Symbol: MIR208B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032786.1 | 1 | 30 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myosin heavy chain 6 | MYH6 | MyHC-alpha | -10.4kb | Sense | Chr. 1 - 163271604 - 163296474 on NCBI Build EquCab3.0 |
Downstream | neuroguidin | NGDN | 45.6kb | Anti-sense | Chr. 1 - 163352503 - 163359626 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
