Assay Details
| Gene Symbol: MIR146B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032778.1 | 1 | 37 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC111773378 | LOC111773378 | Anti-sense | Chr. 1 - 27874058 - 27887206 on NCBI Build EquCab3.0 | ||
| Downstream | CUE domain containing 2 | CUEDC2 | 10.3kb | Anti-sense | Chr. 1 - 27892914 - 27900533 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR146B
Genomic Map