Assay Details
Gene Symbol: MIR598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032808.1 | 1 | 32 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | XK related 6 | XKR6 | Sense | Chr. 2 - 59333319 - 59638957 on NCBI Build EquCab3.0 | ||
Downstream | myotubularin related protein 9 | MTMR9 | 270.6kb | Anti-sense | Chr. 2 - 59727665 - 59787840 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
