Assay Details
| Gene Symbol: MIR95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032816.1 | 1 | 41 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | actin binding LIM protein family member 2 | ABLIM2 | Sense | Chr. 3 - 117492156 - 117649118 on NCBI Build EquCab3.0 | ||
| Downstream | SH3 domain and tetratricopeptide repeats 1 | SH3TC1 | 150.8kb | Anti-sense | Chr. 3 - 117669802 - 117728900 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR95
Genomic Map