Assay Details
| Gene Symbol: MIR1204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032882.1 | 1 | 37 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
MYC proto-oncogene, bHLH transcription factor |
MYC | -42.6kb | Sense | Chr. 9 - 71418587 - 71423659 on NCBI Build EquCab3.0 | |
| Downstream | uncharacterized LOC102149291 | LOC102149291 | 398.5kb | Sense | Chr. 9 - 71864790 - 71960771 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR1204
Genomic Map