Assay Details
Gene Symbol: MIR655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033042.1 | 1 | 42 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-544-2 | MIR544-2 | eca-mir-544-2 | -813.0 | Sense | Chr. 24 - 43357324 - 43357413 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-3959 | MIR3959 | eca-mir-3959 | 1.9kb | Sense | Chr. 24 - 43360172 - 43360282 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
