Assay Details
Gene Symbol: MIR486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033059.1 | 1 | 28 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin 1 | ANK1 | Sense | Chr. 27 - 3513129 - 3729356 on NCBI Build EquCab3.0 | ||
Downstream | NK6 homeobox 3 | NKX6-3 | 10.0kb | Sense | Chr. 27 - 3731577 - 3735614 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
