Assay Details
Gene Symbol: MIR331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033062.1 | 1 | 52 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
vezatin, adherens junctions transmembrane protein |
VEZT | Sense | Chr. 28 - 21377268 - 21457555 on NCBI Build EquCab3.0 | ||
Downstream | methionyl aminopeptidase 2 | METAP2 | 111.1kb | Sense | Chr. 28 - 21561787 - 21598500 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
