Assay Details
| Gene Symbol: MIR545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033109.1 | 1 | 56 | 87 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-374b | MIR374B | eca-mir-374b | -64.3kb | Sense | Chr. X - 58556190 - 58556261 on NCBI Build EquCab3.0 |
| Downstream | microRNA mir-374a | MIR374A | eca-mir-374a | 79.0 | Sense | Chr. X - 58620768 - 58620839 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR545
Genomic Map