Assay Details
Gene Symbol: MIR660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033111.1 | 1 | 55 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-500-2 | MIR500-2 | eca-mir-500-2 | -1.3kb | Sense | Chr. X - 41480016 - 41480099 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-502 | MIR502 | eca-mir-502 | 781.0 | Sense | Chr. X - 41482322 - 41482401 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
