Assay Details
Gene Symbol: MIR199A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_035003.1 | 1 | 37 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-214 | MIR214 | eca-mir-214 | -5.7kb | Sense | Chr. 5 - 8117501 - 8117565 on NCBI Build EquCab3.0 |
Downstream | chromosome 5 C1orf105 homolog | C5H1orf105 | 247.0kb | Anti-sense | Chr. 5 - 8370311 - 8406726 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
