Assay Details
| Gene Symbol: MIR8990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_127996.1 | 1 | 26 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translation initiation factor IF-2-like | LOC111774391 | -29.2kb | Sense | Chr. 7 - 100102002 - 100263384 on NCBI Build EquCab3.0 | |
| Downstream | WT1 transcription factor | WT1 | 675.0 | Sense | Chr. 7 - 100293420 - 100347376 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR8990
Genomic Map