Assay Details
Gene Symbol: MIR9032 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128046.1 | 1 | 48 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glyoxylate and hydroxypyruvate reductase | GRHPR | -3.1kb | Anti-sense | Chr. 25 - 2554107 - 2564411 on NCBI Build EquCab3.0 | |
Downstream | RNA polymerase I subunit E | POLR1E | 49.6kb | Anti-sense | Chr. 25 - 2617194 - 2634188 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
