Assay Details
| Gene Symbol: MIR148B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032853.1 | 1 | 38 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | COPI coat complex subunit zeta 1 | COPZ1 | Sense | Chr. 6 - 72043105 - 72061640 on NCBI Build EquCab3.0 | ||
| Downstream | G protein-coupled receptor 84 | GPR84 | 21.0kb | Anti-sense | Chr. 6 - 72071140 - 72073783 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR148B
Genomic Map