Assay Details
| Gene Symbol: MIR9068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128091.1 | 1 | 125 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Gse1 coiled-coil protein | GSE1 | Anti-sense | Chr. 3 - 32951505 - 33357766 on NCBI Build EquCab3.0 | ||
| Downstream | microRNA mir-9069 | MIR9069 | eca-mir-9069 | 84.4kb | Anti-sense | Chr. 3 - 33231552 - 33231678 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9068
Genomic Map