Assay Details
Gene Symbol: MIR9071 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128094.1 | 1 | 23 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
GrpE like 1, mitochondrial |
GRPEL1 | -2.2kb | Anti-sense | Chr. 3 - 116711314 - 116719041 on NCBI Build EquCab3.0 | |
Downstream | sortilin related VPS10 domain containing receptor 2 | SORCS2 | 99.6kb | Sense | Chr. 3 - 116821036 - 117304910 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
