Assay Details
Gene Symbol: MIR9085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128108.1 | 1 | 65 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin C | CCNC | Sense | Chr. 10 - 51788095 - 51814859 on NCBI Build EquCab3.0 | ||
Downstream | PR/SET domain 13 | PRDM13 | 37.0kb | Anti-sense | Chr. 10 - 51850409 - 51858748 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
