Assay Details
| Gene Symbol: MIR9108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128136.1 | 1 | 58 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 11 | TMEM11 | -17.0kb | Sense | Chr. 11 - 61422854 - 61436220 on NCBI Build EquCab3.0 | |
| Downstream | mitogen-activated protein kinase kinase 3 | MAP2K3 | 30.8kb | Anti-sense | Chr. 11 - 61484160 - 61510874 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9108
Genomic Map