Assay Details
Gene Symbol: MIR9114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128144.1 | 1 | 51 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC111775906 | LOC111775906 | -37.2kb | Sense | Chr. 12 - 12297825 - 12320244 on NCBI Build EquCab3.0 | |
Downstream | receptor-type tyrosine-protein phosphatase eta-like | LOC111775907 | 137.2kb | Sense | Chr. 12 - 12494765 - 12600263 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
