Assay Details
Gene Symbol: MIR9135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128168.1 | 1 | 30 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transport and Golgi organization protein 1 homolog | LOC111767535 | Sense | Chr. 13 - 27980266 - 27998358 on NCBI Build EquCab3.0 | ||
Downstream | transport and Golgi organization protein 1 homolog | LOC111767536 | 11.3kb | Sense | Chr. 13 - 28002764 - 28021017 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
