Assay Details
Gene Symbol: MIR9167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128242.1 | 1 | 123 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | exosome component 7 | EXOSC7 | -46.4kb | Anti-sense | Chr. 16 - 43106588 - 43138487 on NCBI Build EquCab3.0 | |
Downstream | SEM1 26S proteasome complex subunit | SEM1 | SHFM1 | 11.3kb | Sense | Chr. 16 - 43196348 - 43200583 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
