Assay Details
Gene Symbol: MIR8975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127981.1 | 1 | 121 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | D-2-hydroxyglutarate dehydrogenase | D2HGDH | Sense | Chr. 6 - 26956811 - 26986832 on NCBI Build EquCab3.0 | ||
Downstream | galactose-3-O-sulfotransferase 2 | GAL3ST2 | 23.8kb | Sense | Chr. 6 - 26986967 - 27012857 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
