Assay Details
Gene Symbol: MIRLET7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033053.1 | 1 | 37 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-99a-2 | MIR99A-2 | eca-mir-99a-2 | -662.0 | Sense | Chr. 26 - 16008719 - 16008799 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-125b-2 | MIR125B-2 | eca-mir-125b-2 | 43.4kb | Sense | Chr. 26 - 16052911 - 16052980 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
