Assay Details
| Gene Symbol: MIR8951 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_127948.1 | 1 | 64 | 120 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 23 C9orf85 homolog | C23H9orf85 | -24.6kb | Anti-sense | Chr. 23 - 18416714 - 18467340 on NCBI Build EquCab3.0 | |
| Downstream | cell migration inducing hyaluronidase 2 | CEMIP2 | TMEM2 | 107.2kb | Sense | Chr. 23 - 18599325 - 18671383 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR8951
Genomic Map