Assay Details
Gene Symbol: MIR8956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127957.1 | 1 | 96 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MYC associated factor X | MAX | -99.1kb | Anti-sense | Chr. 24 - 11640125 - 11676387 on NCBI Build EquCab3.0 | |
Downstream | fucosyltransferase 8 | FUT8 | 129.2kb | Sense | Chr. 24 - 11904846 - 12182178 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
