Assay Details
| Gene Symbol: MIR8974 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_127980.1 | 1 | 92 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuromedin U receptor 1 | NMUR1 | -38.4kb | Sense | Chr. 6 - 18688246 - 18698511 on NCBI Build EquCab3.0 | |
| Downstream | testis expressed 44 | TEX44 | C6H2orf57 | 2.0kb | Anti-sense | Chr. 6 - 18739024 - 18740396 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR8974
Genomic Map