Assay Details
| Gene Symbol: MIR9035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128049.1 | 1 | 72 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | translation initiation factor IF-2-like | LOC102149769 | -40.0kb | Anti-sense | Chr. 25 - 36734254 - 36737642 on NCBI Build EquCab3.0 | |
| Downstream | uncharacterized LOC111770629 | LOC111770629 | 23.8kb | Sense | Chr. 25 - 36801572 - 36810593 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9035
Genomic Map