Assay Details
| Gene Symbol: MIR9065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128088.1 | 1 | 87 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | AT-hook DNA binding motif containing 1 | AHDC1 | -95.3kb | Sense | Chr. 2 - 28305229 - 28369433 on NCBI Build EquCab3.0 | |
| Downstream | G protein-coupled receptor 3 | GPR3 | 9.1kb | Anti-sense | Chr. 2 - 28474039 - 28483826 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9065
Genomic Map