Assay Details
| Gene Symbol: MIR9069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128092.1 | 1 | 68 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-9068 | MIR9068 | eca-mir-9068 | -84.4kb | Anti-sense | Chr. 3 - 33146997 - 33147141 on NCBI Build EquCab3.0 |
| Downstream | GINS complex subunit 2 | GINS2 | 127.1kb | Anti-sense | Chr. 3 - 33358810 - 33367278 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9069
Genomic Map