Assay Details
Gene Symbol: MIR9095 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128121.1 | 1 | 93 | 116 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | TGFB1-induced anti-apoptotic factor 1 | TIAF1 | -82.0kb | Sense | Chr. 11 - 43257737 - 43258666 on NCBI Build EquCab3.0 | |
Downstream | crystallin beta A1 | CRYBA1 | 52.9kb | Anti-sense | Chr. 11 - 43393724 - 43399698 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
