Assay Details
| Gene Symbol: MIR9111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128140.1 | 1 | 87 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tripartite motif containing 44 | TRIM44 | -126.1kb | Sense | Chr. 12 - 2288587 - 2394433 on NCBI Build EquCab3.0 | |
| Downstream | microRNA mir-9123 | MIR9123 | eca-mir-9123 | 69.7kb | Sense | Chr. 12 - 2590376 - 2590510 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR9111
Genomic Map