Assay Details
Gene Symbol: MIR9129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128161.1 | 1 | 94 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GSG1 like | GSG1L | Sense | Chr. 13 - 21293954 - 21515753 on NCBI Build EquCab3.0 | ||
Downstream | katanin interacting protein | KATNIP | KIAA0556 | 122.6kb | Anti-sense | Chr. 13 - 21517079 - 21719685 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
