Assay Details
Gene Symbol: MIR506A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128195.1 | 1 | 89 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-514a | MIR514A | eca-mir-514a | -4.8kb | Sense | Chr. X - 120848435 - 120848529 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-507a | MIR507A |
MIR507, eca-mir-507, eca-mir-507a |
192.0 | Sense | Chr. X - 120853637 - 120853731 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
