Assay Details
Gene Symbol: MIR9160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128234.1 | 1 | 88 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ferritin heavy chain | LOC100630595 | -32.1kb | Sense | Chr. 14 - 47231735 - 47232473 on NCBI Build EquCab3.0 | |
Downstream | zinc finger protein 608 | ZNF608 | 1.3mb | Anti-sense | Chr. 14 - 48572863 - 48673593 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
