Assay Details
| Gene Symbol: LOC100073305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504043.6 | XP_001504093.1 | 2810 | 77 | ||
| XM_023618242.1 | XP_023474010.1 | 3085 | 77 | |||
| XM_023618243.1 | XP_023474011.1 | 528 | 77 | |||
More Information
Set Membership: |
|

LOC100073305
Genomic Map