Assay Details
| Gene Symbol: CD1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252301.1 | NP_001239230.1 | 1 - 2 | 56 | 89 | |
| GenBank mRNA | JF749874.1 | - | 1 - 2 | 56 | 89 | |
More Information
Set Membership: |
|

CD1E1
Genomic Map