Assay Details
| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256913.1 | NP_001243842.1 | 1 | 1822 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPM2AIP1| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256913.1 | NP_001243842.1 | 1 | 1822 | 83 | |
Set Membership: |
|